SY3H3D682B8A Synthonix Corporation

4-(1-(o-Tolyl)vinyl)benzoic acid

  • Cas Number : 1799412-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/58636.html

Safety Information

MSDS https://synthonix.com/58636.html
Availability details for
Quantity
  • Availability
  • Add to Cart