SY3H3D682B8C Synthonix Corporation

4,4,5,5-Tetramethyl-2-(4-nitronaphthalen-1-yl)-1,3,2-dioxaborolane

  • MDL number No MDL found
  • Cas Number : 1565857-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58709.html

Safety Information

MSDS https://synthonix.com/58709.html
Availability details for
Quantity
  • Availability
  • Add to Cart