SY3H3D682B8D Synthonix Corporation

3,3',5,5'-Tetramethylbiphenyl-4,4'-diol

  • MDL number No MDL found

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/58710.html

Safety Information

MSDS https://synthonix.com/58710.html
Availability details for
Quantity
  • Availability
  • Add to Cart