SY3H3D682B90 Synthonix Corporation

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-1-ol

  • MDL number No MDL found
  • Cas Number : 1560648-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58734.html

Safety Information

MSDS https://synthonix.com/58734.html
Availability details for
Quantity
  • Availability
  • Add to Cart