SY3H3D682B92 Synthonix Corporation

2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine

  • MDL number No MDL found
  • Cas Number : 1214264-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/58740.html

Safety Information

MSDS https://synthonix.com/58740.html
Availability details for
Quantity
  • Availability
  • Add to Cart