SY3H3D682B98 Synthonix Corporation

(1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropyl)methanol

  • MDL number No MDL found
  • Cas Number : 1220219-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/58817.html

Safety Information

MSDS https://synthonix.com/58817.html
Availability details for
Quantity
  • Availability
  • Add to Cart