SY3H3D682B99 Synthonix Corporation

4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine

  • MDL number No MDL found
  • Cas Number : 852227-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/58826.html

Safety Information

MSDS https://synthonix.com/58826.html
Availability details for
Quantity
  • Availability
  • Add to Cart