SY3H3D682BA2 Synthonix Corporation

3-((4,5,6,7-Tetrahydro-1H-indol-2-yl)methylene)indolin-2-one

  • MDL number MFCD14586259
  • Cas Number : 204003-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58926.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58926.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart