SY3H3D682BA3 Synthonix Corporation

2,2',3,3'-Tetrahydro-1,1'-spirobi[indene]-7,7'-diol

  • Cas Number : 223137-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/58940.html

Safety Information

MSDS https://synthonix.com/58940.html
Availability details for
Quantity
  • Availability
  • Add to Cart