SY3H3D682BAE Synthonix Corporation

Tris(3-fluorophenyl)phosphine

  • Cas Number : 23039-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59133.html

Safety Information

MSDS https://synthonix.com/59133.html
Availability details for
Quantity
  • Availability
  • Add to Cart