SY3H3D682BB2 Synthonix Corporation

Tris(4-iodophenyl)amine

  • MDL number No MDL found
  • Cas Number : 4181-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59148.html

Safety Information

MSDS https://synthonix.com/59148.html
Availability details for
Quantity
  • Availability
  • Add to Cart