SY3H3D682BB6 Synthonix Corporation

[1,1':3',1''-Terphenyl]-5'-ylboronic acid

  • Cas Number : 128388-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/59184.html

Safety Information

MSDS https://synthonix.com/59184.html
Availability details for
Quantity
  • Availability
  • Add to Cart