SY3H3D682BB8 Synthonix Corporation

1,1,3-Trimethyl-3-phenyl-2,3-dihydro-1H-indene

  • MDL number No MDL found
  • Cas Number : 3910-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/59229.html

Safety Information

MSDS https://synthonix.com/59229.html
Availability details for
Quantity
  • Availability
  • Add to Cart