SY3H3D682BB9 Synthonix Corporation

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole

  • MDL number No MDL found
  • Cas Number : 855738-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/59234.html

Safety Information

MSDS https://synthonix.com/59234.html
Availability details for
Quantity
  • Availability
  • Add to Cart