SY3H3D682BBC Synthonix Corporation

4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-2-ol

  • Cas Number : 1219741-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59253.html

Safety Information

MSDS https://synthonix.com/59253.html
Availability details for
Quantity
  • Availability
  • Add to Cart