SY3H3D682BBD Synthonix Corporation

2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9a-dihydro-4aH-carbazole

  • MDL number No MDL found
  • Cas Number : 871125-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/59261.html

Safety Information

MSDS https://synthonix.com/59261.html
Availability details for
Quantity
  • Availability
  • Add to Cart