SY3H3D682BBF Synthonix Corporation

4,4,5,5-Tetramethyl-2-(1,2,3,6-tetrahydro-[1,1'-biphenyl]-4-yl)-1,3,2-dioxaborolane

  • MDL number No MDL found
  • Cas Number : 287944-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59291.html

Safety Information

MSDS https://synthonix.com/59291.html
Availability details for
Quantity
  • Availability
  • Add to Cart