SY3H3D682BC0 Synthonix Corporation

1-((2,4,6-Triisopropylphenyl)sulfonyl)-1H-imidazole

  • MDL number No MDL found
  • Cas Number : 50257-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59296.html

Safety Information

MSDS https://synthonix.com/59296.html
Availability details for
Quantity
  • Availability
  • Add to Cart