SY3H3D682BC1 Synthonix Corporation

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-tosyl-1,2,3,6-tetrahydropyridine

  • MDL number No MDL found
  • Cas Number : 1256360-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/59305.html

Safety Information

MSDS https://synthonix.com/59305.html
Availability details for
Quantity
  • Availability
  • Add to Cart