SY3H3D682C0E Synthonix Corporation

((3S,5R)-5-((1H-1,2,4-Triazol-1-yl)methyl)-5-(2,4-difluorophenyl)tetrahydrofuran-3-yl)methyl 4-chlorobenzenesulfonate

  • MDL number No MDL found
  • Cas Number : 175712-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59512.html

Safety Information

MSDS https://synthonix.com/59512.html
Availability details for
Quantity
  • Availability
  • Add to Cart