SY3H3D682C10 Synthonix Corporation

5,5,10,10-Tetramethyl-5,10-dihydroindeno[2,1-a]indene

  • Cas Number : 89057-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/59532.html

Safety Information

MSDS https://synthonix.com/59532.html
Availability details for
Quantity
  • Availability
  • Add to Cart