SY3H3D682C14 Synthonix Corporation

Undecane-1,11-diol

  • MDL number No MDL found
  • Cas Number : 765-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/55159.html

Safety Information

MSDS https://synthonix.com/55159.html
Availability details for
Quantity
  • Availability
  • Add to Cart