SY3H3D682C23 Synthonix Corporation

6-Vinylnaphthalen-2-ol

  • MDL number No MDL found
  • Cas Number : 136896-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55606.html

Safety Information

MSDS https://synthonix.com/55606.html
Availability details for
Quantity
  • Availability
  • Add to Cart