SY3H3D682C24 Synthonix Corporation

1-(4-Vinylbenzyl)-1H-imidazole

  • MDL number No MDL found
  • Cas Number : 78430-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55700.html

Safety Information

MSDS https://synthonix.com/55700.html
Availability details for
Quantity
  • Availability
  • Add to Cart