SY3H3D682C30 Synthonix Corporation

Oxazol-2(3H)-one

  • MDL number No MDL found
  • Cas Number : 27584-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/39828.html

Safety Information

MSDS https://synthonix.com/39828.html
Availability details for
Quantity
  • Availability
  • Add to Cart