SY3H3D682C39 Synthonix Corporation

Oxazolidine-4-carboxylic acid hydrochloride

  • MDL number No MDL found
  • Cas Number : 1219384-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/40338.html

Safety Information

MSDS https://synthonix.com/40338.html
Availability details for
Quantity
  • Availability
  • Add to Cart