SY3H3D682C47 Synthonix Corporation

Oxazolo[4,5-b]pyridine

  • MDL number MFCD13175183
  • Cas Number : 273-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42083.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42083.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart