SY3H3D682C49 Synthonix Corporation

Oxazolo[5,4-b]pyridine

  • MDL number No MDL found
  • Cas Number : 273-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42085.html

Safety Information

MSDS https://synthonix.com/42085.html
Availability details for
Quantity
  • Availability
  • Add to Cart