SY3H3D682C4D Synthonix Corporation

Oxazolo[5,4-c]pyridin-2-amine

  • MDL number No MDL found
  • Cas Number : 1260890-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42471.html

Safety Information

MSDS https://synthonix.com/42471.html
Availability details for
Quantity
  • Availability
  • Add to Cart