SY3H3D682C50 Synthonix Corporation

6-Oxo-1,6-dihydropyridine-3-carboxylic acid hydrazide

  • Cas Number : 134531-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/43112.html

Safety Information

MSDS https://synthonix.com/43112.html
Availability details for
Quantity
  • Availability
  • Add to Cart