SY3H3D682C9C Synthonix Corporation

3-(4-Oxo-2-thioxothiazolidin-3-yl)propanoic acid

  • MDL number No MDL found
  • Cas Number : 7025-19-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43171.html

Safety Information

MSDS https://synthonix.com/43171.html
Availability details for
Quantity
  • Availability
  • Add to Cart