SY3H3D682CA0 Synthonix Corporation

2-Oxopyrrolidin-3-yl acetate

  • MDL number No MDL found
  • Cas Number : 42491-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43385.html

Safety Information

MSDS https://synthonix.com/43385.html
Availability details for
Quantity
  • Availability
  • Add to Cart