SY3H3D682CA1 Synthonix Corporation

(1S,4S)-2-Oxa-5-azabicyclo[2.2.1]heptan-3-one methanesulfonate

  • MDL number No MDL found
  • Cas Number : 769167-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/43582.html

Safety Information

MSDS https://synthonix.com/43582.html
Availability details for
Quantity
  • Availability
  • Add to Cart