SY3H3D682CA4 Synthonix Corporation

3-Oxocyclohexanecarboxylic acid

  • Cas Number : 16205-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43724.html

Safety Information

MSDS https://synthonix.com/43724.html
Availability details for
Quantity
  • Availability
  • Add to Cart