SY3H3D682CA7 Synthonix Corporation

3-Oxa-1,8-diazaspiro[4.5]decan-2-one

  • MDL number No MDL found
  • Cas Number : 945947-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/43794.html

Safety Information

MSDS https://synthonix.com/43794.html
Availability details for
Quantity
  • Availability
  • Add to Cart