SY3H3D682CA8 Synthonix Corporation

Octahydro-1H-pyrrolo[3,4-b]pyridine

  • Cas Number : 5654-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43870.html

Safety Information

MSDS https://synthonix.com/43870.html
Availability details for
Quantity
  • Availability
  • Add to Cart