SY3H3D682CAC Synthonix Corporation

4-Oxo-3,4-dihydrothieno[2,3-d]pyrimidine-5-carbonitrile

  • MDL number No MDL found
  • Cas Number : 1801143-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/44087.html

Safety Information

MSDS https://synthonix.com/44087.html
Availability details for
Quantity
  • Availability
  • Add to Cart