SY3H3D682CAD Synthonix Corporation

5-OXO-4,5-DIHYDROPYRAZOLO[1,5-A]PYRIMIDINE-3-CARBONITRILE

  • MDL number No MDL found
  • Cas Number : 1224288-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44409.html

Safety Information

MSDS https://synthonix.com/44409.html
Availability details for
Quantity
  • Availability
  • Add to Cart