SY3H3D682CB3 Synthonix Corporation

4-Oxocyclohexyl acetate

  • MDL number No MDL found
  • Cas Number : 41043-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47022.html

Safety Information

MSDS https://synthonix.com/47022.html
Availability details for
Quantity
  • Availability
  • Add to Cart