SY3H3D682CC3 Synthonix Corporation

(1S,3aR,6aS)-Octahydrocyclopenta[c]pyrrole-1-carboxylic acid hydrochloride

  • MDL number No MDL found
  • Cas Number : 1205676-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49553.html

Safety Information

MSDS https://synthonix.com/49553.html
Availability details for
Quantity
  • Availability
  • Add to Cart