SY3H3D682CC4 Synthonix Corporation

1-Oxa-3,9-diazaspiro[5.5]undecan-2-one hydrochloride

  • MDL number No MDL found
  • Cas Number : 1598381-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49556.html

Safety Information

MSDS https://synthonix.com/49556.html
Availability details for
Quantity
  • Availability
  • Add to Cart