SY3H3D682CC5 Synthonix Corporation

cis-Octahydro-1H-isoindole hydrochloride

  • Cas Number : 161829-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49562.html

Safety Information

MSDS https://synthonix.com/49562.html
Availability details for
Quantity
  • Availability
  • Add to Cart