SY3H3D682CC6 Synthonix Corporation

6-oxa-2-Azaspiro[4.5]decane hydrochloride

  • MDL number MFCD28894400
  • Cas Number : 1956322-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49564.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49564.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart