SY3H3D682CCA Synthonix Corporation

Octahydro-2H-chromen-2-one

  • MDL number No MDL found
  • Cas Number : 4430-31-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50435.html

Safety Information

MSDS https://synthonix.com/50435.html
Availability details for
Quantity
  • Availability
  • Add to Cart