SY3H3D682CCD Synthonix Corporation

(3aS,7aS)-Octahydro-1H-indole-2-carboxylic acid

  • Cas Number : 2055118-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50469.html

Safety Information

MSDS https://synthonix.com/50469.html
Availability details for
Quantity
  • Availability
  • Add to Cart