SY3H3D682CCF Synthonix Corporation

3-Oxoisoindoline-5-carbonitrile

  • MDL number No MDL found
  • Cas Number : 1261726-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/50809.html

Safety Information

MSDS https://synthonix.com/50809.html
Availability details for
Quantity
  • Availability
  • Add to Cart