SY3H3D682CD2 Synthonix Corporation

4-Oxo-3,4-dihydroquinazoline-7-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 202197-73-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50829.html

Safety Information

MSDS https://synthonix.com/50829.html
Availability details for
Quantity
  • Availability
  • Add to Cart