SY3H3D682CD4 Synthonix Corporation

4-Oxo-1,4-dihydroquinazoline-6-carboxamide

  • Cas Number : 150454-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51145.html

Safety Information

MSDS https://synthonix.com/51145.html
Availability details for
Quantity
  • Availability
  • Add to Cart