SY3H3D682CD5 Synthonix Corporation

2-Oxoindoline-3-carbaldehyde

  • Cas Number : 78610-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51176.html

Safety Information

MSDS https://synthonix.com/51176.html
Availability details for
Quantity
  • Availability
  • Add to Cart