SY3H3D682CD6 Synthonix Corporation

3-Oxoisoindoline-5-carbaldehyde

  • MDL number No MDL found
  • Cas Number : 1260664-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51184.html

Safety Information

MSDS https://synthonix.com/51184.html
Availability details for
Quantity
  • Availability
  • Add to Cart